missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human IFN-gamma/IL-2 Dual-Color ELISpot Kit

highly sensitive, microplate-based assays for the detection of cytokine secreting cells

Brand:  R&D Systems™ ELD4506

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13465135

  • £456.32 / 1 set


Estimated Shipment
03-04-2019
Add to basket

Includes:
  • Microplate
  • Biotin-conjugated Detection Antibody
  • HRP-conjugated Detection Antibody
  • Streptavidin-conjugated to Alkaline Phosphatase
  • Dilution Buffers
  • Wash Buffer Concentrate
  • BCIP/NBT Chromogen
  • AEC

Description

Description

Product Features:
  • Detect and quantitate individual cells secreting IFN-γ/IL-2
  • High sensitivity - ELISpot assays can measure responses with frequencies well below 1 in 100,000 cells
  • No in vitro expansion of cells required
  • High-throughput - ELISpot assays use only a small number of primary cells
Specifications

Specifications

IFN-gamma/IL-2
Detection of cytokine secreting cells, quantitative sandwich ELISA
Human IFN-gamma/IL-2 Dual-Color ELISpot Kit
Store the unopened product at 2 - 8° C. Do not use past expiration date.
Human
Human IFN-gamma/IL-2 Microplate, Biotin-conjugated Detection Antibody, HRP-conjugated Detection Antibody, Streptavidin-conjugated to Alkaline Phosphatase, Dilution Buffers, Wash Buffer Concentrate, BCIP/NBT Chromogen, AEC Chromogen, Human IFN-gamma/IL-2 Positive Controls
Whole cells
1 x 96-well plate
Videos