Learn More
R&D Systems™ Human IFN-gamma/IL-17 Dual-Color ELISpot Kit
highly sensitive, microplate-based assays for the detection of cytokine secreting cells
Brand:  R&D Systems™ ELD5219
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13445145
  • £515.97 / 1 set
Call for latest availability
+44 (0)1509 555 500
Specifications
Specifications
Diagnostics
Human IFNG/IL-17 Dual-Color ELISpot Kit
One 96-well plate
One 96-well plate coated w/mAb for human IFNG and pAb for human IL-17, Biotin- and HRP-conjugated detection Abs, Streptavidin-AP, Buffers, BCIP/NBT, AEC, Positive controls
Store the unopened product at 2 - 8° C. Do not use past expiration date.