missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human IFN-gamma Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 285-IF-100

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13441954

  • £357.72 / 100µg

Estimated Shipment date
from Supplier 23-09-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Human IFN-gamma Protein is derived from E. coli. The Recombinant Human IFN-gamma Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in Sodium Succinate, Mannitol and Tween™ 80 with BSA as a carrier protein.
16.9 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Unlabeled
3458
IFN-gamma
100μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes
Videos
Documents