missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human IFN-alpha A, Carrier-Free Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
POA
Specifications
Formulation | Phosphate buffered saline (PBS) |
---|---|
Purification Method | A combination of ion exchange, hydrophobic interaction and size exclusion chromatography |
Recombinant | Yes |
Description
The Recombinant Human IFN-alpha A Protein is derived from E. coli. The Recombinant Human IFN-alpha A Protein has been validated for the following applications: Bioactivity.Specifications
Phosphate buffered saline (PBS) | |
A combination of ion exchange, hydrophobic interaction and size exclusion chromatography | |
Yes |