missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human IFN-alpha A, Carrier-Free Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Brand:  R&D Systems™ 11101-2

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13438553

  • £266.65 / 20µg

Estimated Shipment
09-07-2019

Please to purchase this item. Need a web account? Register with us today!


Specifications

Specifications

Phosphate buffered saline (PBS)
>95%
Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
A combination of ion exchange, hydrophobic interaction and size exclusion chromatography
20μg
Yes
Videos