missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human HVEM/TNFRSF14 Fc Chimera Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 356-HV-100

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13443214

  • £244.21 / 100µg

Estimated Shipment
27-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Source: Mouse myeloma cell line, NS0-derived, Pro37-Val202 (Ser108Thr and Ala140Arg)
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 45 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 8764
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
45 kDa (monomer)
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Unlabeled
8764
HVEM/TNFRSF14 Fc Chimera
100μg
Disulfide-linked homodimer
Yes
Videos