Learn More
R&D Systems™ Human His6 UBE2R2 Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity
Brand:  R&D Systems™ E2-715-100
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13434955
  • £296.94 / 100µg
Estimated Shipment date
from Supplier 02-04-2020
Description
Description
The Recombinant Human His6-UBE2R2 Protein is derived from E. coli. The Recombinant Human His6-UBE2R2 Protein has been validated for the following applications: Enzyme Activity.
Specifications
Specifications
Q712K3
His6 UBE2R2
Human
Unlabeled
54926
100μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening.
Yes