missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human His6 UBE2Q1 Isoform 1 Recombinant Protein

Brand:  R&D Systems™ E2-755-100

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13464965

  • £258.73 / 100µg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Source: E. coli-derived, Contains an N-terminal 6-His tag
Entrez Gene ID: 55585
Regulatory Status: RUO

Specifications

Specifications

Q7Z7E8
His6 UBE2Q1 Isoform 1
Human
Yes
55585
100μg
Unlabeled
Videos