Learn More
R&D Systems™ Human His6 UBE2J1 Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand:  R&D Systems™ E2-750-100
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13444965
  • £288.12 / 100µg
Estimated Shipment date
from Supplier 21-11-2019
Description
Description
The Recombinant Human His6-UBE2J1 Protein is derived from E. coli. The Recombinant Human His6-UBE2J1 Protein has been validated for the following applications: Enzyme Activity.
Specifications
Specifications
Q9Y385
51465
100μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening.
Yes
X mg/mL (X μM) in 50 mM HEPES pH 8.0, 200 mM NaCl, 10% Glycerol (v/v), 1 mM TCEP
His6 UBE2J1
Human
Unlabeled