missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human His6 UBE2F Recombinant Protein

Brand:  R&D Systems™ E2-679-100

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13464935

  • £258.73 / 100µg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Source: E. coli-derived, Contains an N-terminal 6-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Formulation: Supplied as a solution in HEPES, NaCl, DTT and Glycerol.
Entrez Gene ID: 140739
Regulatory Status: RUO

Specifications

Specifications

Supplied as a solution in HEPES, NaCl, DTT and Glycerol.
His6 UBE2F
100μg
Unlabeled
140739
95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Human
Yes
Videos