missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human HGF Recombinant Protein, (NS0-expressed)

Brand:  R&D Systems™ 294-HGN-005/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13462114

  • £249.49 / 5µg


Estimated Shipment
07-03-2019
Add to basket

Description

Description

  • Source: Mouse myeloma cell line, NS0-derived, Gln32-Ser728
  • Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
  • Molecular Weight: alpha chain: 53.7 kDabeta chain: 26 kDa
  • Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
  • Entrez Gene ID: 3082
  • Regulatory Status: RUO
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
alpha chain: 53.7 kDabeta chain: 26 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Unlabeled
3082
HGF
5μg
Disulfide-linked heterodimer
Yes
Videos