missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human HGF R/c-MET Fc Chimera Recombinant Protein

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity

Brand:  R&D Systems™ 358-MT-100/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13443254

  • £260.05 / 100µg

Estimated Shipment
01-07-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Human HGF R/c-MET Fc Chimera His-tag Protein is derived from NS0. The Recombinant Human HGF R/c-MET Fc Chimera His-tag Protein has been validated for the following applications: Binding Activity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
32.5 kDa (alpha chain), 96.7 kDa (beta chain)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
NS0
Tetramer containing two disulfide-linked heterodimers (proteolytically cleaved alpha and beta subunits)
Yes
4233
HGF R/c-MET Fc Chimera
100μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unlabeled
Videos