Learn More
R&D Systems™ Human GST UBE2M/Ubc12 Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand:  R&D Systems™ E2-655-100
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13444915
  • £288.12 / 100µg
Estimated Shipment date
from Supplier 18-12-2019
Description
Description
The Recombinant Human GST-UBE2M/Ubc12 Protein is derived from E. coli. The Recombinant Human GST-UBE2M/Ubc12 Protein has been validated for the following applications: Enzyme Activity.
Specifications
Specifications
No Formulation Info in Bulk Inventory
GST UBE2M/Ubc12
100μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening.
Yes
9040
95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Human
Unlabeled