missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human GST UbcH8 Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Brand:  R&D Systems™ E2-643-100

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13444905

  • £258.73 / 100µg

Estimated Shipment
27-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Human GST-UbcH8/UBE2L6 Protein is derived from E. coli. The Recombinant Human GST-UbcH8/UBE2L6 Protein has been validated for the following applications: Enzyme Activity.
Specifications

Specifications

No Formulation Info in Bulk Inventory
GST UbcH8
100μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening.
Yes
9246
95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Human
Unlabeled
Videos