Learn More
R&D Systems™ Human GST UbcH5a/UBE2D1 Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity
Brand:  R&D Systems™ E2-615-100
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13434875
  • £296.94 / 100µg
Estimated Shipment date
from Supplier 06-04-2020
Description
Description
The Recombinant Human GST-UbcH5a/UBE2D1 Protein is derived from E. coli. The Recombinant Human GST-UbcH5a/UBE2D1 Protein has been validated for the following applications: Enzyme Activity.
Specifications
Specifications
X mg/mL (X μM) in 50 mM HEPES pH 8.0, 200 mM NaCl, 10% Glycerol (v/v), 1 mM TCEP
44 kDa
95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Human
Unlabeled
7321
GST UbcH5a/UBE2D1
100μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening.
Yes