missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Growth Hormone Recombinant Protein

Brand:  R&D Systems™ 1067-GH-025

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13458433

  • £149.85 / 25µg


Estimated Shipment
04-03-2019
Add to basket

Description

Description

Source: E. coli-derived, Phe27-Phe217, with an N-terminal Met
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 22 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Tris and NaCl.
Entrez Gene ID: 2688
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in Tris and NaCl with BSA as a carrier protein.
22 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
2688
Growth Hormone
25μg
Unlabeled
Videos