missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Growth Hormone 2 Recombinant Protein

Brand:  R&D Systems™ 7668-GH-025

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13486274

  • £312.00 / 25µg


Estimated Shipment
05-07-2018
Add to basket

Description

Description

Source: E. coli-derived, Phe27-Phe217, with an N-terminal Met
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 22.4 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in HCl.
Entrez Gene ID: 2689
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein.
22.4 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
2689
Growth Hormone 2
25μg
Unlabeled
Videos