missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Glypican 5 Recombinant Protein

Brand:  R&D Systems™ 2607-G5-050

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13461564

  • £245.53 / 50µg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Source: Mouse myeloma cell line, NS0-derived, Glu25-Thr554, with a C-terminal 6-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 59.9 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 2262
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
59.9 kDa (monomer)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
2262
Glypican 5
50μg
Unlabeled
Videos