missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human GITR Ligand/TNFSF18 Protein, CF
A New and Improved rh GITR-L is Available! CHO expressed; ∼70 fold better activity; HA tag!
POA
Specifications
Conjugate | Unconjugated |
---|---|
Gene ID (Entrez) | 8995 |
Name | GITR Ligand/TNFSF18 |
Quantity | 25 µg |
Recombinant | Yes |
Description
Measured by its ability to compete with biotinylated recombinant human GITR Ligand for binding to immobilized recombinant human GITR Fc Chimera.Specifications
Unconjugated | |
GITR Ligand/TNFSF18 | |
Yes | |
M.W. (Observed): 14 kDa-16 kDa, reducing conditions; M.W. (theoretical): 15 kDa |
8995 | |
25 µg | |
Spodoptera frugiperda, Sf 21 (baculovirus)-derived human GITR Ligand/TNFSF18 protein Glu74-Ser199, with an N-terminal 6-His tag |