missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human GITR Ligand/TNFSF18 Recombinant Protein, HA tag, (CHO-expressed)

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Brand:  R&D Systems™ 6987-GL-025/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13466014

  • £290.08 / 25µg

Estimated Shipment
17-09-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Human GITR Ligand/TNFSF18 (CHO-expressed) is derived from CHO. The Recombinant Human GITR Ligand/TNFSF18 (CHO-expressed) has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
19.6 kDa (monomer)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Chinese Hamster Ovary cell line,CHO-derived human GITR Ligand/TNFSF18 protein, (N-terminus) Hemagglutinin Tag (YPYDVPDYA) GCN4-IZ GGGSGGGSGGGS Human GITR Ligand (Glu74-Ser199) Accession &Num; NP_005083 (C-terminus)
Non-covalently-linked trimer
Unconjugated
8995
GITR Ligand/TNFSF18
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Activation-inducible TNF-related ligand, AITRL, AITRLMGC138237, GITRL, GITRLglucocorticoid-induced TNFR-related protein ligand, Glucocorticoid-induced TNF-related ligand, hGITRLGITR ligand, TL6AITR ligand, TNFSF18, tumor necrosis factor (ligand) superfamily, member 18, tumor necrosis factor ligand superfamily member 18
Yes
Videos
Documents