missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human GDF-15 Recombinant Protein

Brand:  R&D Systems™ 957-GD-025/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13486444

  • £359.00 / 25µg


Estimated Shipment
05-07-2018
Add to basket

Description

Description

Source: Chinese Hamster Ovary cell line, CHO-derived, Ala197-Ile308, with an N-terminal 6-His tag
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 13 kDa (monomer). Based on mass spectrometric analysis, a carboxyl-terminal truncated peptide with a molecular mass of approximately 12.5 kDa is also present in the recombinant protein preparation.
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 9518
Regulatory Status: RUO

Specifications

Specifications

Reconstitute at 100-300μg/mL in sterile 4mM HCI.
9518
GDF-15
25μg
Disulfide-linked homodimer
Yes
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Predicted MW: 13kDa (monomer) Observed MW: 14kDa, reducing conditions
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Source: Chinese Hamster Ovary cell line, CHO-derived Ala197-Ile308, with an N-terminal 6-His tag; N Terminal Sequence: His & Asn199
Unlabeled
Videos