missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human GDF-15 Recombinant Protein

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Brand:  R&D Systems™ 957-GD-025

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13466444

  • £407.19 / 25µg

Estimated Shipment date
from Supplier 25-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Human GDF-15 (CHO-expressed) Protein is derived from CHO.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
Predicted MW: 13kDa (monomer) Observed MW: 14kDa, reducing conditions
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Source: Chinese Hamster Ovary cell line, CHO-derived Ala197-Ile308, with an N-terminal 6-His tag; N Terminal Sequence: His & Asn199
Disulfide-linked homodimer
Yes
9518
GDF-15
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unlabeled
Videos
Documents