missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human GASP-1/WFIKKNRP Recombinant Protein

Brand:  R&D Systems™ 2070-GS-025/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13460414

  • £245.53 / 25µg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Source: Mouse myeloma cell line, NS0-derived, Leu35-His576, with a C-terminal 6-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 60.7 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 124857
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
60.7 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
124857
GASP-1/WFIKKNRP
25μg
Unlabeled
Videos