missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Galectin-3 Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 1154-GA-050

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13448753

  • £393.96 / 50µg

Estimated Shipment date
from Supplier 24-01-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Human Galectin-3 Protein is derived from E. coli. The Recombinant Human Galectin-3 Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS and Betamercaptoethanol with BSA as a carrier protein.
26 kDa
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Unlabeled
3958
Galectin-3
50μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 3 months, 2 to 8° C under sterile conditions after reconstitution.
Yes
Videos
Documents