missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Galectin-3 Recombinant Protein

Brand:  R&D Systems™ 1154-GA-050/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13458753

  • £343.21 / 50µg


Estimated Shipment
06-03-2019
Add to basket

Description

Description

Source: E. coli-derived, Ala2-Ile250
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 26 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS and Betamercaptoethanol.
Entrez Gene ID: 3958
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS and Betamercaptoethanol.
26 kDa
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
3958
Galectin-3
50μg
Unlabeled
Videos