missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Galectin-1 Recombinant Protein

Brand:  R&D Systems™ 1152-GA-050

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13488743

  • £331.00 / 50µg


Estimated Shipment
02-07-2018
Add to basket

Description

Description

Source: E. coli-derived, Ala2-Asp135
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 15 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS and Betamercaptoethanol.
Entrez Gene ID: 3956
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS and Betamercaptoethanol with BSA as a carrier protein.
15 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
3956
Galectin-1
50μg
Unlabeled
Videos