missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human G-CSF Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 214-CS-025/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13430674

  • £598.29 / 25µg

Estimated Shipment date
from Supplier 02-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Human G-CSF Protein is derived from E. coli. The Recombinant Human G-CSF Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Supplied as a 0.2μm filtered solution in Acetic Acid.
18.8 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Unlabeled
1440
G-CSF
25μg
Do not freeze. 6 months from date of receipt, 2 to 8° C as supplied.
Yes
Videos
Documents