missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human G-CSF R/CD114 Recombinant Protein

Brand:  R&D Systems™ 381-GR-050/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13463654

  • £340.57 / 50µg


Estimated Shipment
03-09-2018
Add to basket

Description

Description

Source: Mouse myeloma cell line, NS0-derived, Glu25-Pro621
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 66.5 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 1441
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
66.5 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
1441
G-CSF R/CD114
50μg
Unlabeled
Videos