missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human FSH alpha/beta Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 5925-FS-010

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13455604

  • £371.91 / 10µg

Estimated Shipment date
from Supplier 06-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Human FSH alpha/beta Protein is derived from CHO. The Recombinant Human FSH alpha/beta Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
10.2 kDa (FSH alpha) & 12.4 kDa (FSH beta)
95%, by SDS-PAGE with silver staining.
Chinese Hamster Ovary cell line, CHO-derived Human FSH alpha Ala25-Ser116 Accession # P01215 Human FSH beta Cys21-Glu129 Accession # P01225
Noncovalently-linked heterodimer
Yes
2488
FSH alpha/beta
10μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unlabeled
Videos
Documents