missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Follistatin 315 aa 30-344 Recombinant Protein

Brand:  R&D Systems™ 4889-FN-025

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13465154

  • £360.37 / 25µg


Estimated Shipment
05-04-2019
Add to basket

Description

Description

Measured by its ability to neutralize Activin-mediated erythroid differentiation of K562 human chronic myelogenous leukemia cells. The ED 50 for this effect is 0.01-0.05 µg/mL in the presence of 7.5 ng/mL of rhActivin A.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with Trehalose and with BSA as a carrier protein.
34.7 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Unlabeled
10468
Follistatin 315 aa 30-344
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes
Videos