missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Follistatin 315 aa 30-344 Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
POA
Specifications
Conjugate | Unconjugated |
---|---|
Gene ID (Entrez) | 10468 |
Molecular Weight (g/mol) | 34.7 kDa |
Name | Follistatin 315 aa 30-344 |
Quantity | 25μg |
Description
The Recombinant Human Follistatin (aa 30-344) Protein is derived from CHO. The Recombinant Human Follistatin (aa 30-344) Protein has been validated for the following applications: Bioactivity.Specifications
Unconjugated | |
34.7 kDa | |
25μg | |
Chinese Hamster Ovary cell line,CHO-derived human Follistatin protein Gly30-Trp344 |
10468 | |
Follistatin 315 aa 30-344 | |
Yes |