missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Flt-3 Ligand Recombinant Protein, (NSO derived)

Brand:  R&D Systems™ 308-FKN-005

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13462374

  • £163.68 / 5µg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Source: Mouse myeloma cell line, NS0-derived, Thr27-Pro185
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 18 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 2323
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
18 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
2323
Flt-3 Ligand
5μg
Unlabeled
Videos