missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human FGF basic 146 aa Recombinant Protein

Brand:  R&D Systems™ 233-FB-025/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13461064

  • £209.88 / 25µg


Estimated Shipment
01-03-2019
Add to basket

Description

Description

Source: E. coli-derived, Pro143-Ser288, with an N-terminal Ala
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 16.5 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Tris-HCl and NaCl.
Entrez Gene ID: 2247
Regulatory Status: RUO

Specifications

Specifications

Reconstitute at 100-250μg/mL in sterile PBS.
2247
FGF basic 146 aa
25μg
Unlabeled
Lyophilized from a 0.2μm filtered solution in Tris-HCl and NaCl.
16.5 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
Videos