missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human FGF acidic aa 16-155 Recombinant Protein

Brand:  R&D Systems™ 232-FA-025/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13461044

  • £195.36 / 25µg


Estimated Shipment
25-02-2019
Add to basket

Description

Description

Source: E. coli-derived, Phe16-Asp155, with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 15.5 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in MOPS, Na2SO4 and EDTA.
Entrez Gene ID: 2246
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in MOPS, Na2SO4 and EDTA.
15.5 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
2246
FGF acidic aa 16-155
25μg
Unlabeled
Videos