missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human FGF-5 Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 237-F5-050/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13431114

  • £185.22 / 50µg

Estimated Shipment date
from Supplier 31-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Human FGF-5 Protein is derived from E. coli. The Recombinant Human FGF-5 Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Supplied as a 0.2 μm filtered solution in MOPS, Na 2 SO 4 EDTA and DTT.
27 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Unlabeled
2250
FGF-5
50μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after opening.
Yes
Videos
Documents