missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human FGF-19 Recombinant Protein

Brand:  R&D Systems™ 969-FG-025/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13486464

  • £278.00 / 25µg


Estimated Shipment
02-07-2018
Add to basket

Description

Description

Source: E. coli-derived, Leu25-Lys216
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 21 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 9965
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
21 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
9965
FGF-19
25μg
Unlabeled
Videos