missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human/Feline/Rhesus Macaque CXCL12/SDF-1 alpha Recombinant Protein

Brand:  R&D Systems™ 350-NS-010/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13463104

  • £249.49 / 10µg


Estimated Shipment
04-03-2019
Add to basket

Description

Description

Source: E. coli-derived, Lys22-Lys89
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 8.0 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 6387
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
8.0 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human/Feline/Rhesus Macaque
Yes
6387
CXCL12/SDF-1 alpha
10μg
Unlabeled
Videos