missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Fc gamma RIIIB/CD16b Recombinant Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity
POA
Specifications
Conjugate | Unconjugated |
---|---|
Gene ID (Entrez) | 2215 |
Molecular Weight (g/mol) | 22.7 kDa |
Name | Fc gamma RIIIB/CD16b |
Quantity | 50μg |
Description
The Recombinant Human Fc gamma RIIIB/CD16b Protein is derived from NS0. The Recombinant Human Fc gamma RIIIB/CD16b Protein has been validated for the following applications: Binding Activity.Specifications
Unconjugated | |
22.7 kDa | |
50μg | |
Mouse myeloma cell line,NS0-derived human Fc gamma RIIIB/CD16b protein Thr20-Gln208,with a C-terminal 10-His tag |
2215 | |
Fc gamma RIIIB/CD16b | |
Yes |