missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Fc gamma RIIIB/CD16b Recombinant Protein

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity

Brand:  R&D Systems™ 1597-FC-050/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13439763

  • £398.37 / 50µg

Estimated Shipment date
from Supplier 16-12-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Human Fc gamma RIIIB/CD16b Protein is derived from NS0. The Recombinant Human Fc gamma RIIIB/CD16b Protein has been validated for the following applications: Binding Activity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
22.7 kDa
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Unlabeled
2215
Fc gamma RIIIB/CD16b
50μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes
Videos
Documents