missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Fas/TNFRSF6/CD95 Fc Chimera Recombinant Protein, 97%

Brand:  R&D Systems™ 326-FS-050

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13462694

  • £264.01 / 50µg


Estimated Shipment
28-02-2019
Add to basket

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
45 kDa (monomer)
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Unlabeled
355
Fas/TNFRSF6/CD95 Fc Chimera
50μg
Disulfide-linked homodimer
Yes
Videos