missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Fas Ligand/TNFSF6 Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

POA

Specifications

Conjugate Unconjugated
Gene ID (Entrez) 356
Molecular Weight 18 kDa
Name Fas Ligand/TNFSF6
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs

Description

Description

The Recombinant Human Fas Ligand/TNFSF6 Protein is derived from CHO. The Recombinant Human Fas Ligand/TNFSF6 Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Unconjugated
18 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
356
Fas Ligand/TNFSF6
10μg
Chinese Hamster Ovary cell line,CHO-derived human Fas Ligand/TNFSF6 protein Pro134-Leu281,with an N-terminal Met and 6-His tag