missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Epiregulin Recombinant Protein

Brand:  R&D Systems™ 1195-EP-025

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13488833

  • £233.65 / 25µg


Estimated Shipment
04-10-2018
Add to basket

Description

Description

Source: E. coli-derived, Val63-Leu108, with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 5.4 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 2069
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
5.4 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Unlabeled
2069
Epiregulin
25μg
<0.1 EU per 1μg of the protein by the LAL method.
Yes
Videos