missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Epiregulin Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
POA
Specifications
Gene ID (Entrez) | 2069 |
---|---|
Molecular Weight (g/mol) | 5.4 kDa |
Name | Epiregulin |
Quantity | 25μg |
Source | E. coli-derived human Epiregulin protein Val63-Leu108,with an N-terminal Met |
Description
The Recombinant Human Epiregulin Protein is derived from E. coli. The Recombinant Human Epiregulin Protein has been validated for the following applications: Bioactivity.Specifications
2069 | |
Epiregulin | |
E. coli-derived human Epiregulin protein Val63-Leu108,with an N-terminal Met | |
<0.1 EU per 1μg of the protein by the LAL method. | |
Recombinant |
5.4 kDa | |
25μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |