missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Dkk-2 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
POA
Specifications
For Use With (Application) | Bioactivity |
---|---|
Gene ID (Entrez) | 27123 |
Molecular Weight (g/mol) | 25.8 kDa |
Name | Dkk-2 |
Quantity | 10μg |
Description
The Recombinant Human Dkk-2 Protein is derived from CHO. The Recombinant Human Dkk-2 Protein has been validated for the following applications: Bioactivity.Specifications
Bioactivity | |
25.8 kDa | |
10μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
27123 | |
Dkk-2 | |
Chinese Hamster Ovary cell line,CHO-derived human Dkk-2 protein Met1-Ile259 | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |