missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human CXCL9/MIG Recombinant Protein

Brand:  R&D Systems™ 392-MG-010

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13463824

  • £168.96 / 10µg


Estimated Shipment
04-04-2019
Add to basket

Description

Description

Measured by its ability to chemoattract BaF3 mouse pro-B cells transfected with mouse CXCR3. The ED 50 for this effect is 0.1-0.4 µg/mL.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
11.7 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Unlabeled
4283
CXCL9/MIG
10μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes
Videos