missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human CXCL9/MIG Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
POA
Specifications
Conjugate | Unconjugated |
---|---|
Gene ID (Entrez) | 4283 |
Molecular Weight (g/mol) | 11.7 kDa |
Name | CXCL9/MIG |
Recombinant | Yes |
Description
The Recombinant Human CXCL9/MIG Protein is derived from E. coli. The Recombinant Human CXCL9/MIG Protein has been validated for the following applications: Bioactivity.Specifications
Unconjugated | |
11.7 kDa | |
Yes |
4283 | |
CXCL9/MIG | |
E. coli-derived human CXCL9/MIG protein Thr23-Thr125 |