missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human CXCL8/IL-8 Recombinant Protein

Brand:  R&D Systems™ 208-IL-010/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13460454

  • £171.60 / 10µg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Source: E. coli-derived, Ser28-Ser99
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 8 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in deionized water.
Entrez Gene ID: 3576
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in deionized water.
8 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
3576
CXCL8/IL-8
10μg
Unlabeled
Videos