missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human CXCL8/IL-8 Mucin Stalk Chimera Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 968-IL-025/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13466464

  • £244.02 / 25µg

Estimated Shipment date
from Supplier 25-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Human IL-8/CXCL8 (Mucin Stalk Chimera) is derived from NS0. The Recombinant Human IL-8/CXCL8 (Mucin Stalk Chimera) has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
34.8 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse myeloma cell line, NS0-derived Human IL-8 (Ala23-Ser99) (Asn98Gly) & (Ser28-Ser99) (Asn98Gly) Accession # P10145 Human Fractalkine Mucin-like Stalk (Phe103-Gln341) Accession # P78423 KQN 6-His tag
3-10C, AMCF-I, C-X-C motif chemokine 8, CXCL8, CXCL8SCYB8, Emoctakin, GCP1, GCP-1TSG-1, IL8, interleukin 8, K60, LAI, LECT, LUCT, LYNAP, MDNCF, MDNCFb-ENAP, member 8, MONAP, MONAPGCP1, NAF, NAP1, NAP-1NAP1, NCF, Neutrophil-activating protein 1, Protein 3-10C, T cell chemotactic factor, T-cell chemotactic factor, TCF, TSG1
Yes
3576
CXCL8/IL-8 Mucin Stalk Chimera
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unlabeled
Videos
Documents