missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human CXCL3/GRO gamma Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
POA
Specifications
For Use With (Application) | Bioactivity |
---|---|
Gene ID (Entrez) | 2921 |
Molecular Weight (g/mol) | 8 kDa |
Name | CXCL3/GRO gamma/CINC-2/DCIP-1 |
Source | E. coli-derived human CXCL3/GRO gamma/CINC-2/DCIP-1 protein Ala35-Asn107 |
Description
The Recombinant Human CXCL3/GRO gamma Protein is derived from E. coli. The Recombinant Human CXCL3/GRO gamma Protein has been validated for the following applications: Bioactivity.Specifications
Bioactivity | |
8 kDa | |
E. coli-derived human CXCL3/GRO gamma/CINC-2/DCIP-1 protein Ala35-Asn107 | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
2921 | |
CXCL3/GRO gamma/CINC-2/DCIP-1 | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |