missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human CXCL3/GRO gamma/CINC-2/DCIP-1 Recombinant Protein

Brand:  R&D Systems™ 277-GG-050/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13460584

  • £295.69 / 50µg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Source: E. coli-derived, Ala35-Asn107
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 8 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 2921
Regulatory Status: RUO

Specifications

Specifications

Bioactivity
2921
CXCL3/GRO gamma/CINC-2/DCIP-1
50μg
Unlabeled
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
8 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
Videos